Tetramethylammonium chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TMACl, TMA

13833-1000 AC13833-0010 AC13833-2500
TS13833-1000EA 48 USD
TS13833-1000 TS13833-0010 TS13833-2500
Tetramethylammonium chloride 98+%
Tetramethylammonium chloride
Formula: (CH₃)₄NCl
Melting Pt: 420 °C
Storage Temperature: Ambient
MDL Number: MFCD00011628
CAS Number: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.0 %
Stabilizer ≤0.05 % tert-Butylcatechol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR