Tetraheptylammonium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

21816-1000 21816-0250
TS21816-1000EA 432 USD
TS21816-1000 TS21816-0250
Tetraheptylammonium bromide 99%
Tetraheptylammonium bromide
Formula: C₂₈H₆₀BrN
MW: 490.7 g/mol
Melting Pt: 96
Storage Temperature: Ambient
MDL Number: MFCD00011861
CAS Number: 4368-51-8
EINECS: 224-459-3

Order Now

Specification Test Results

Appearance White Flakes
Infrared spectrum Conforms
Melting point 89 to 91 °C
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR