Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
42012-1000
42012-5000
420120025
TS42012-1000EA
145
USD
TS42012-1000
TS42012-5000
TS420120025
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Tetrabutylammonium hydroxide
Formula:
C₁₆H₃₇NO MW: 259.48 g/mol Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009425 CAS Number: 2052-49-5 EINECS: 218-147-6 UN: 3267 ADR: 8,II |
Specification Test Results
Appearance | Clear solution |
UV | at 420 nm A: ≤0.15 |
Color scale | ≤10 APHA |
Titration with HCl | 0.37 to 0.43 M |
Bromide (Br) | ≤500 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...