Tetrabutylammonium bromide 99+%

Supplier: Thermo Scientific Chemicals
Warning

18568-1000 AC18568-0025 AC18568-0250 AC18568-0500 AC18568-5000
TS18568-1000EA 96 USD
TS18568-1000 TS18568-0025 TS18568-0250 TS18568-0500 TS18568-5000
Tetrabutylammonium bromide 99+%
Tetrabutylammonium bromide
Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
Water ≤1 %
HPLC ≥94 % (sum of tylosins A+B+C+D)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR