Octadecyltrimethoxysilane 90% tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethoxy(octadecyl)silane

31360-0250 AC31360-1000
TS31360-0250EA 124 USD
TS31360-0250 TS31360-1000
Octadecyltrimethoxysilane 90% tech.
Octadecyltrimethoxysilane
Formula: C₂₁H₄₆O₃Si
MW: 374.68 g/mol
Boiling Pt: 170 °C (.1 mmHg)
Melting Pt: 13…17 °C
Density: 0.88 g/cm³
Flash Pt: 139 °C
Storage Temperature: Ambient
MDL Number: MFCD00043060
CAS Number: 3069-42-9
EINECS: 221-339-2

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to light yellow Liquid (May develop some turbidity or precipitate. May partially crystallize)
Infrared spectrum Conforms
GC ≥88 % Total octadecyltrimethoxysilanes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR