N,N-Dimethylformamide 99.9%, AcroSeal™ for biochemistry

Supplier: Thermo Scientific Chemicals
Danger

44709-0010 447090025
TS44709-0010EA 147.06 USD
TS44709-0010 TS447090025
N,N-Dimethylformamide 99.9%, AcroSeal™ for biochemistry
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (760 mmHg)
Melting Pt: –61 °C
Density: 0.945 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.85 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR