N,N-Dimethylformamide ≥99.8% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

42364-0010 423640025 423640100 423640250 423645000
TS42364-0010EA 131 USD
TS42364-0010 TS423640025 TS423640100 TS423640250 TS423645000
N,N-Dimethylformamide ≥99.8% ACS
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: -61
Density: 0.945 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.15 % (coulometric)
GC ≥99.8 %
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %
Titrable acid ≤0.0005 meq/g
Titrable base ≤0.003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR