Magnesium oxide 98%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesia usta

20515-5000 AC20515-0025
TS20515-5000EA 56 USD
TS20515-5000 TS20515-0025
Magnesium oxide 98%, pure
Magnesium oxide
Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: 2800 °C
Density: 3.58 g/cm³
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specification Test Results

Appearance White to gray Crystalline powder
Titration Complexometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR