Magnesium carbonate, meets analytical specification of USP for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium Carbonate

43913-0010 AC43913-2500
TS43913-0010EA 273 USD
TS43913-0010 TS43913-2500
Magnesium carbonate, meets analytical specification of USP for biochemistry
Magnesium carbonate
Formula: CMgO₃
MW: 84.31
Density: 3.03
Storage Temperature: Ambient
MDL Number: MFCD00064632
CAS Number: 546-93-0

Order Now

Specification Test Results

Appearance White Powder
Additional info Absence-specified microorganisms: Passes
Identification Conforms
Titration excess HCl 40.0 to 43.5 % (MgO)
Arsenic (As) ≤4 ppm
Calcium (Ca) ≤0.45 %
Iron (Fe) ≤0.02 %
Acid insoluble substances ≤0.05 %
Soluble salts ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR