Cetrimonium bromide 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
22716-0025
22716-5000
22716-0100
22716-0101
22716-0250
22716-1000
TS22716-0025EA
823
USD
TS22716-0025
TS22716-5000
TS22716-0100
TS22716-0101
TS22716-0250
TS22716-1000
Cetrimonium bromide 99+%
Cetrimonium bromide
Formula:
CH₃(CH₂)₁₅N(Br)(CH₃)₃ Melting Pt: 230 °C Flash Pt: 244 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011772 CAS Number: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9,III Merck Index: 13,02034 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...