D-(+)-Trehalose, anhydrous 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (+)-Trehalose, α,α-Trehalose, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol), Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside, D-(+)-Trehalose Anhydrous, D-Trehalose

30987-0250 30987-1000 30987-5000
TS30987-0250EA 151 USD
TS30987-0250 TS30987-1000 TS30987-5000
D-(+)-Trehalose, anhydrous 99%
D-(+)-Trehalose
Formula: C₁₂H₂₂O₁₁
Melting Pt: 203 °C
Storage Temperature: Ambient
MDL Number: MFCD00006628
CAS Number: 99-20-7
EINECS: 202-739-6
Merck Index: 12,09713

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
Sulfated ash ≤0.1 % (on dry basis)
Solubility (10 % in water) Clear colorless to light yellow
HPLC ≥98.5 % (ELSD) on dry basis
Specific optical rotation +194° to +200° (20°C, 589 nm) (c=7, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR