Chromium(III) chloride hexahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chromium trichloride hexahydrate, Chromic chloride hexahydrate
21336-5000
AC21336-0051
TS21336-5000EA
81
USD
TS21336-5000
TS21336-0051
Chromium(III) chloride hexahydrate 98%
Chromium(III) chloride hexahydrate
Formula:
CrCl₃·6H₂O MW: 266.45 g/mol Melting Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149660 CAS Number: 10060-12-5 EINECS: 233-038-3 UN: 3260 ADR: 8,III Merck Index: 13,02242 |
Specification Test Results
Appearance | Dark green Crystalline powder |
Titration with Na2S2O3 | ≥97.5 % |
Sulfate (SO4) | ≤500 ppm |
Iron (Fe) | ≤300 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...