Chlorotrimethylsilane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TMCS, TMSC, Trimethylchlorosilane

AC11012-0010 AC11012-0250 AC11012-1000 AC11012-2500
TS11012-0025EA 203.39 USD
TS11012-0025 TS11012-0010 TS11012-0250 TS11012-1000 TS11012-2500
Chlorotrimethylsilane 98%
Chlorotrimethylsilane
Formula: C₃H₉ClSi
MW: 108.64 g/mol
Boiling Pt: 57 °C (760 mmHg)
Melting Pt: –58 °C
Density: 0.85 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000502
CAS Number: 75-77-4
EINECS: 200-900-5
UN: 1298
ADR: 3,II
REACH: 01-2119457596-25

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3890 (20°C, 589 nm)
GC ≥97.5 %
Enantiomeric excess ≥97.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR