Calcium sulfate dihydrate 98+% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Plaster of Paris

31525-5000 AC31525-0025
TS31525-5000EA 168 USD
TS31525-5000 TS31525-0025
Calcium sulfate dihydrate 98+% ACS
Calcium sulphate dihydrate
Formula: CaSO₄·2H₂O
MW: 172.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric 98.0 to 102.0 %
Chloride (Cl) ≤50 ppm
Carbonate (CO3) Passes test
Nitrate (NO3) (limit about 0.003%) passes test
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤200 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤200 ppm
Strontium (Sr) ≤500 ppm
Heavy metals (ICP-OES) ≤0.002 %
Insoluble matter ≤0.02 % (in dilute HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR