Calcium bromide hydrate, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
45063-0025
AC45063-2500
TS45063-0025EA
321
USD
TS45063-0025
TS45063-2500
Calcium bromide hydrate, extra pure
Calcium bromide hydrate
Formula:
Br₂Ca·H₂O MW: 217.9 g/mol Melting Pt: 730 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149608 CAS Number: 71626-99-8 |
Specification Test Results
Appearance | White Crystalline powder and/or chunks |
Titration Complexometric | ≥82 % (CaBr2) |
Solubility | (10 % in water) Clear colorless to slightly hazy |
Acidity/alkalinity | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...