Bromoacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

10657-2500 106570010 106570025 106570100
TS10657-2500EA 74 USD
TS10657-2500 TS106570010 TS106570025 TS106570100
Bromoacetic acid 98+%
Bromoacetic acid
Formula: BrCH₂COOH
Boiling Pt: 208 °C (760 mmHg)
Melting Pt: 49…51 °C
Density: 1.93 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002678
CAS Number: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: 8,II
Merck Index: 13,01384

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals or flakes or mass and/or chunks
Infrared spectrum Conforms
Melting point 45 to 49 °C
Titration with NaOH 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR