Ammonium sulphide 20% in water

Supplier: Thermo Scientific Chemicals
Danger

42608-1000 426080010
TS42608-1000EA 80 USD
TS42608-1000 TS426080010
Ammonium sulphide 20% in water
Ammonium sulphide
Formula: H₈N₂S
MW: 68.14
Melting Pt: -18
Density: 0.99
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00010892
CAS Number: 12135-76-1
UN: 2683
ADR: 8 II
Merck Index: 13,00560

Order Now

Specification Test Results

Appearance Clear yellow Solution
Titration Iodimetric 20.0 to 24.0 %
Refractive index 1.5290 to 1.5320 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR