p-Toluenesulphonyl chloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Toluene-4-sulphonyl chloride, Tosyl chloride, 4-Toluenesulphonyl chloride, Toluene-p-sulphonyl chloride

13903-1000 13903-0020 13903-0100 13903-0250 13903-5000
TS13903-1000EA 59 USD
TS13903-1000 TS13903-0020 TS13903-0100 TS13903-0250 TS13903-5000
p-Toluenesulphonyl chloride 99+%
p-Toluenesulphonyl chloride
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 265
Melting Pt: 67
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00007450
CAS Number: 98-59-9
EINECS: 202-684-8
UN: 3261
ADR: 8 II
Merck Index: 13,09612

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 69 °C
Assay ≥99.0 %
Impurity ≤0.3 % Total free acid (as toluenesulfonic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR