4-Cyanophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

35904-0100 35904-0010
TS35904-0100EA 332 USD
TS35904-0100 TS35904-0010
4-Cyanophenylboronic acid 98%
4-Cyanophenylboronic acid
Formula: C₇H₆BNO₂
MW: 146.94 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –106 °C
Density: 0.72 g/cm³
Flash Pt: 2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318968
CAS Number: 126747-14-6
EINECS: 203-486-4
UN: 2050
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR