Allyl alcohol 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

10286-0025 10286-0010 10286-2500
TS10286-0025EA 123 USD
TS10286-0025 TS10286-0010 TS10286-2500
Allyl alcohol 99%, Extra Pure
Allyl alcohol
Formula: C₃H₆O
Boiling Pt: 96…98 °C (760 mmHg)
Melting Pt: –129 °C
Density: 0.85 g/cm³
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00002920
CAS Number: 107-18-6
EINECS: 203-470-7
UN: 1098
ADR: 6.1,
Merck Index: 13,00283

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4115 to 1.4145 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥98.5 %
Color scale ≤20 APHA
Acidity (CH3COOH) ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR