2-Methyl-2-butanol 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Amyl alcohol, tert-Pentyl alcohol, tert-Pentanol

16662-0010 166620025 166620100
TS16662-0010EA 166 USD
TS16662-0010 TS166620025 TS166620100
2-Methyl-2-butanol 99%, pure
2-Methyl-2-butanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 102 °C (760 mmHg)
Melting Pt: –12 °C
Density: 0.8 g/cm³
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00004478
CAS Number: 75-85-4
EINECS: 200-908-9
UN: 1105
ADR: 3,II
Merck Index: 13,07218

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
Water ≤0.1 %
GC ≥98.5 %
Sulfate (SO4) ≤10 ppm
Silicate (as SiO2) ≤5 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.5 ppm
Nickel (Ni) ≤0.1 ppm
Nitrogen (N) ≤10 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.5 ppm
Zinc (Zn) ≤0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR