Diisopropyl ether 98+% stabilized, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
15896-0010
158960025
158960100
TS15896-0010EA
91
USD
TS15896-0010
TS158960025
TS158960100
Diisopropyl ether 98+% stabilized, pure
Diisopropyl ether
Formula:
(CH₃)₂CHOCH(CH₃)₂ Boiling Pt: 68 °C (760 mmHg) Melting Pt: –85.5 °C Density: 0.72 g/cm³ Flash Pt: –29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008880 CAS Number: 108-20-3 EINECS: 203-560-6 UN: 1159 ADR: 3,II Merck Index: 13,05232 |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3670 to 1.3690 (20°C, 589 nm) |
Water | ≤0.5 % |
GC | ≥98.0 % |
Color scale | ≤15 APHA |
Stabilizer | 5 to 150 ppm BHT |
Acidity | ≤0.005 % |
Peroxides | ≤0.002 % (as H2O2) |
Mercury (Hg) | ≤0.0001 % |
Zinc (Zn) | ≤0.001 % |
Heavy metals (as Pb) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...