N-Ethyldiisopropylamine 99.5+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA

45959-0010 459591000
TS45959-0010EA 653 USD
TS45959-0010 TS45959-1000 TS459591000
N-Ethyldiisopropylamine 99.5+%, AcroSeal™
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 127
Melting Pt: -50
Density: 0.756 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
UN: 3384
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4150 (20°C, nm)
Water ≤0.3 %
GC ≥99.50 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR