11-Bromo-1-undecanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 11-Bromoundecan-1-ol, 11-Bromoundecanol

29344-0500 29344-0100
TS29344-0500EA 196 USD
TS29344-0500 TS29344-0100
11-Bromo-1-undecanol 97%
11-Bromo-1-undecanol
Formula: HO(CH₂)₁₀CH₂Br
MW: 251.21 g/mol
Boiling Pt: 165…170 °C (1 mmHg)
Melting Pt: 42…47 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004752
CAS Number: 1611-56-9
EINECS: 216-554-3

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR