1-Octanesulfonic acid sodium salt monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate

20614-0250 20614-1000 AC20614-0050
TS20614-0250EA 207 USD
TS20614-0250 TS20614-1000 TS20614-0050
1-Octanesulfonic acid sodium salt monohydrate 98%
1-Octanesulphonic acid sodium salt monohydrate
Formula: C₈H₁₇NaO₃S·1H₂O
MW: 234.29 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 207596-29-0
EINECS: 226-195-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying 7 % to 9 % (140°C)
Titration after Ion exchange ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR