3-Pentyn-1-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pent-3-yn-1-ol

19733-1000 19733-0010 19733-0050 19733-0250
TS19733-1000EA 1084 USD
TS19733-1000 TS19733-0010 TS19733-0050 TS19733-0250
3-Pentyn-1-ol 99%
3-Pentyn-1-ol
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 154…157 °C (760 mmHg)
Density: 0.91 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00002956
CAS Number: 10229-10-4
EINECS: 233-550-7
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR