m-Cresol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene

11058-0010 11058-1000 11058-0025 11058-0100 11058-5000
TS11058-0010EA 190 USD
TS11058-0010 TS11058-1000 TS11058-0025 TS11058-0100 TS11058-5000
m-Cresol 99%
m-Cresol
Formula: CH₃C₆H₄OH
MW: 108.14 g/mol
Boiling Pt: 203 °C (760 mmHg)
Melting Pt: 8…10 °C
Density: 1.03 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00002302
CAS Number: 108-39-4
EINECS: 203-577-9
UN: 2076
ADR: 6.1,II
Merck Index: 13,02604
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce465c84cc8b7c230526b22e6f830300b605d109&vid=72c08ae3da371936f9349196418a240331cb79ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light red Viscous liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5420 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR