L(+)-Potassium sodium tartrate tetrahydrate ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

44722-0025 AC44722-1000 AC44722-5000
TS44722-0025EA 233 USD
TS44722-0025 TS44722-1000 TS44722-5000
L(+)-Potassium sodium tartrate tetrahydrate ACS
L(+)-Potassium sodium tartrate tetrahydrate
Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Melting Pt: 70…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
pH 6.0 to 8.5 (5% soln. at 25°C)
Titration with HClO4 99.0 to 102.0 %
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤0.002 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.002 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR