Benzyl alcohol 99%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phenylmethanol
, Benzenemethanol
, Phenylcarbinol
148390250
14839-0010
148390025
TS148390025EA
196
USD
TS148390025
TS148390250
TS14839-0010
Benzyl alcohol 99%, pure
Benzyl alcohol
Formula:
C₆H₅CH₂OH MW: 108.14 g/mol Boiling Pt: 205 °C (760 mmHg) Melting Pt: –15 °C Density: 1.044 g/cm³ Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 EINECS: 202-859-9 Merck Index: 13,01126 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5380 to 1.5410 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...