trans-Stilbene 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-1,2-Diphenylethylene, (E)-1,2-Diphenylethene, trans-1,2-Diphenylethene

16104-5000 16104-1000
TS16104-5000EA 463 USD
TS16104-5000 TS16104-1000
trans-Stilbene 96%
trans-Stilbene
Formula: C₁₄H₁₂
Boiling Pt: 366 °C (760 mmHg)
Melting Pt: 25…35 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00064300
CAS Number: 103-30-0
EINECS: 220-744-1
UN: 3335
ADR: 9,III
Merck Index: 12,08972
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52acf17325a810eecda8f5a4d795a5b31a1f6434&vid=6d95185bf3c5a4ec46f2df78329eb0b669b2b255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
Melting point 25 to 35 °C
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR