Nickel(II) sulphate hexahydrate 98+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickelous sulphate, Hexahydrate, Nickel sulphate hexahydrate
41561-0010
41561-5000
AC41561-0051
AC41561-1000
TS41561-0010EA
287
USD
TS41561-0010
TS41561-5000
TS41561-0051
TS41561-1000
Nickel(II) sulphate hexahydrate 98+% ACS
Nickel(II) sulphate hexahydrate
Formula:
NiSO₄·6H₂O MW: 262.85 g/mol Melting Pt: 840 Storage Temperature: Ambient |
MDL Number:
MFCD00149813 CAS Number: 10101-97-0 EINECS: 232-104-9 UN: 3077 ADR: 9 III Merck Index: 13,06541 |
Specification Test Results
Appearance | Blue-green Crystals |
Titration Complexometric | 98.0 to 102.0 % |
Chloride (Cl) | ≤0.001 % |
Calcium (Ca) | ≤0.005 % |
Cobalt (Co) | ≤0.002 % |
Copper (Cu) | ≤0.005 % |
Iron (Fe) | ≤0.001 % |
Magnesium (Mg) | ≤0.005 % |
Manganese (Mn) | ≤0.002 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.05 % |
Insoluble matter | ≤0.005 % |
Nitrogen compounds (as N) | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...