Calcium sulfate, Drierite™ 6 mesh, without indicator

Supplier: Thermo Scientific Chemicals

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum

21904-0100 219040020 219045000
TS21904-0100EA 385 USD
TS21904-0100 TS219040020 TS219045000
Calcium sulfate, Drierite™ 6 mesh, without indicator
Calcium sulphate
Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1450 °C
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Appearance White to grayish-white Irregular granules
Water ≤0.3 %
Particle size 6 mesh
Composition ≥99.5 % (CaSO4) Typical

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR