Sodium-1-decanesulphonate, anhydrous 99+% for ion pair chromatography

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

39671-1000 AC39671-0250
TS39671-1000EA 1137 USD
TS39671-1000 TS39671-0250
Sodium-1-decanesulphonate, anhydrous 99+% for ion pair chromatography
Sodium-1-decanesulphonate
Formula: C₁₀H₂₁NaO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (150°C)
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.2 M soln.) Passes test
Suitability (HPLC gradient) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR