Sodium-1-decanesulphonate 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

36945-1000 AC36945-0050 AC36945-0250
TS36945-1000EA 815 USD
TS36945-1000 TS36945-0050 TS36945-0250
Sodium-1-decanesulphonate 96%
Sodium-1-decanesulphonate
Formula: C₁₀H₂₁NaO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 % (100°C, 3 h) (vacuum)
Titration after Ion exchange ≥95 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR