Potassium-tert-butoxide 20% (w/w) 1.6-1.7 M in THF, AcroSeal™, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
42879-1000
42879-8000
TS42879-1000EA
170
USD
TS42879-1000
TS42879-8000
Potassium-tert-butoxide 20% (w/w) 1.6-1.7 M in THF, AcroSeal™, pure
Potassium-tert-butoxide
Formula:
C₄H₉KO Flash Pt: –21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012162 CAS Number: 865-47-4 UN: 2920 ADR: 8,I |
Specification Test Results
Appearance | Light yellow to amber Solution |
Strength | 1.60 to 1.75 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...