Potassium-tert-butoxide 1 M in THF, AcroSeal™, pure

Supplier: Thermo Scientific Chemicals
Danger

37122-0010 37122-1000 371220025
TS37122-0010EA 460 USD
TS37122-0010 TS37122-1000 TS371220025
Potassium-tert-butoxide 1 M in THF, AcroSeal™, pure
Potassium-tert-butoxide
Formula: C₄H₉KO
Density: 0.902 g/cm³
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Colorless to yellow to light orange Solution (May develop some turbidity or precipitate)
Strength 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR