Calcium hydroxide 95+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Slaked lime, Calcium dihydroxide

40385-0010 40385-0025
TS40385-0010EA 106 USD
TS40385-0010 TS40385-0025
Calcium hydroxide 95+% ACS
Calcium hydroxide
Formula: Ca(OH)₂
MW: 74.09 g/mol
Boiling Pt: 2850 °C
Melting Pt: 550 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
Merck Index: 13,01675

Order Now

Specification Test Results

Appearance White Powder
Titration with HCl ≤3.0 % (CaCO3)
Chloride (Cl) ≤300 ppm
Iron (Fe) ≤500 ppm
Magnesium (Mg) ≤5000 ppm
Potassium (K) ≤500 ppm
Sodium (Na) ≤500 ppm
Strontium (Sr) ≤500 ppm
Heavy metals (ICP-OES) ≤30 ppm
Sulfur compounds (as SO4) ≤0.1 %
Insoluble matter ≤0.03 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR