Iron(III) nitrate nonahydrate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

20249-0050 AC20249-0010 AC20249-2500
TS20249-0050EA 955 USD
TS20249-0050 TS20249-0010 TS20249-2500
Iron(III) nitrate nonahydrate 99+% for analysis
Iron(III) nitrate nonahydrate
Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Melting Pt: 47 °C
Density: 1.68 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1,III
Merck Index: 13,04057

Order Now

Specification Test Results

Appearance Light purple Adhering crystalline powder
Titration Iodometric ≥99.0 %
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤50 ppm
Arsenic (As) ≤0.5 ppm
Calcium (Ca) ≤50 ppm
Copper (Cu) ≤50 ppm
Lead (Pb) ≤10 ppm
Manganese (Mn) ≤200 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤50 ppm
Zinc (Zn) ≤10 ppm
Free acid ≤0.3 % (HNO3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR