Copper(II) nitrate trihydrate 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric nitrate trihydrate, Copper dinitrate trihydrate

45396-0010 AC45396-0050
TS45396-0010EA 85 USD
TS45396-0010 TS45396-0050
Copper(II) nitrate trihydrate 99%, pure
Copper(II) nitrate trihydrate
Formula: Cu(NO₃)₂·3H₂O
MW: 241.6 g/mol
Melting Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00149671
CAS Number: 10031-43-3
UN: 3085
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Dark blue Crystalline powder or crystals
Titration Iodometric 98.5 to 104 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR