Copper(II) nitrate trihydrate 99% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric nitrate trihydrate, Copper dinitrate trihydrate

20768-1000 AC20768-0025 AC20768-5000
TS20768-1000EA 44 USD
TS20768-1000 TS20768-0025 TS20768-5000
Copper(II) nitrate trihydrate 99% for analysis
Copper(II) nitrate trihydrate
Formula: Cu(NO₃)₂·3H₂O
MW: 241.6 g/mol
Melting Pt: 114
Storage Temperature: Ambient
MDL Number: MFCD00149671
CAS Number: 10031-43-3
EINECS: 221-838-5
UN: 3085
ADR: 5.1 II
Merck Index: 13,02671

Order Now

Specification Test Results

Appearance Blue Powder or crystals and/or chunks
Titration Iodometric 98.5 to 104.0 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Zinc (Zn) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR