Hydrazinium sulphate ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrazine sulphate salt, Hydrazine sulphate
42377-0500
42377-1000
42377-5000
TS42377-0500EA
60
USD
TS42377-0500
TS42377-1000
TS42377-5000
Hydrazinium sulphate ACS
Hydrazinium sulphate
Formula:
NH₂NH₂·H₂SO₄ Melting Pt: 254 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044873 CAS Number: 10034-93-2 EINECS: 233-110-4 UN: 3288 ADR: 6.1,III Merck Index: 14,04772 |
|
Specification Test Results
Appearance | White Crystalline powder |
Titration Iodimetric | ≥99.0 % |
Residue after ignition | ≤0.05 % |
Chloride (Cl) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...