Calcium hydride ∼ 93%, grains Ø < 2 mm, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Calcium dihydride
19990-5000
AC19990-0025
AC19990-1000
TS19990-5000EA
386
USD
TS19990-5000
TS19990-0025
TS19990-1000
Calcium hydride ∼ 93%, grains Ø < 2 mm, extra pure
Calcium hydride
Formula:
H₂Ca MW: 42.09 g/mol Melting Pt: 816 Storage Temperature: Ambient |
MDL Number:
MFCD00010897 CAS Number: 7789-78-8 EINECS: 232-189-2 UN: 1404 ADR: 4.3 I Merck Index: 13,01674 |
Specification Test Results
Appearance | Gray Powder |
Assay | ≥92 % (Total Ca) |
Particle size | ≤2 mm |
Chloride (Cl) | ≤5000 ppm |
Aluminium (Al) | ≤100 ppm |
Hydrogen (H) | ≥980 ml/g CaH2 |
Iron (Fe) | ≤100 ppm |
Magnesium (Mg) | ≤8000 ppm |
Nitrogen (N) | ≤2000 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...