1,1,1,3,3,3-Hexafluoro-2-propanol 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP
29341-0100
29341-0500
TS29341-0100EA
136
USD
TS29341-0100
TS29341-0500
1,1,1,3,3,3-Hexafluoro-2-propanol 99.9% for spectroscopy
1,1,1,3,3,3-Hexafluoro-2-propanol
Formula:
C₃H₂F₆O Boiling Pt: 59 °C (760 mmHg) Melting Pt: –4 °C Density: 1.596 g/cm³ Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011651 CAS Number: 920-66-1 EINECS: 213-059-4 UN: 3265 ADR: 8,I Merck Index: 13,09598 |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.01 % |
GC | ≥99.85 % |
UV | at 240 nm A: ≤0.009 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...