di-Ammonium oxalate monohydrate 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

42336-5000 AC42336-0025 AC42336-1000
TS42336-5000EA 152 USD
TS42336-5000 TS42336-0025 TS42336-1000
di-Ammonium oxalate monohydrate 99+% ACS
di-Ammonium oxalate monohydrate
Formula: (NH₄)₂C₂O₄·H₂O
MW: 142.11 g/mol
Boiling Pt: 70 °C (1013 hPa)
Melting Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00149694
CAS Number: 6009-70-7
EINECS: 214-202-3
Merck Index: 13,00541

Order Now

Specification Test Results

Appearance White Crystalline powder
Residue after ignition ≤0.02 %
Titration with KMnO4 99.0 to 101.0 %
Chloride (Cl) ≤0.002 %
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR