N,N-Dimethylacetamide, extra dry over Molecular Sieve 99.5%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
37523-1000
375230010
TS37523-1000EA
79
USD
TS37523-1000
TS375230010
N,N-Dimethylacetamide, extra dry over Molecular Sieve 99.5%, AcroSeal™
N,N-Dimethylacetamide
Formula:
CH₃CON(CH₃)₂ MW: 87.12 g/mol Boiling Pt: 166.1 Melting Pt: -20 Density: 0.937 g/cm³ Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008686 CAS Number: 127-19-5 EINECS: 204-826-4 Merck Index: 13,03254 |
|
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...