Benzophenone 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

10556-0010 10556-0050 10556-5000
TS10556-0010EA 70 USD
TS10556-0010 TS10556-0050 TS10556-5000
Benzophenone 99%, pure
Benzophenone
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305 °C (760 mmHg)
Melting Pt: 47…49 °C
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
Merck Index: 13,01099

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 47 to 49 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR