Imidazole 99+%, crystalline

Supplier: Thermo Scientific Chemicals
Danger

30187-0010 30187-0025 30187-2500
TS30187-0010EA 271 USD
TS30187-0010 TS30187-0025 TS30187-2500
Imidazole 99+%, crystalline
Imidazole
Formula: C₃H₄N₂
Boiling Pt: 255…256 °C (760 mmHg)
Melting Pt: 86…90 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8,III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 86 to 90 °C
Water ≤1 % (K.F.)
GC ≥99.0 %
Titration with HClO4 ≥99.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR