Tricyclohexylphosphine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42161-0050
42161-0010
42161-0250
TS42161-0050EA
93
USD
TS42161-0050
TS42161-0010
TS42161-0250
Tricyclohexylphosphine 97%
Tricyclohexylphosphine
Formula:
C₁₈H₃₃P Melting Pt: 77…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 EINECS: 220-069-2 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 77 to 83 °C |
NMR (31P-NMR) | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...