1,2,3,4,5-Pentamethylcyclopentadiene 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentamethylcyclopentadiene, 1,2,3,4,5-Pentamethylcyclopenta-1,3-diene, 1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene

32083-0250 32083-0010 32083-0050
TS32083-0250EA 511 USD
TS32083-0250 TS32083-0010 TS32083-0050
1,2,3,4,5-Pentamethylcyclopentadiene 98+%
1,2,3,4,5-Pentamethylcyclopentadiene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 68 °C (1013 hPa)
Melting Pt: –98.5 °C
Density: 0.66 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00001354
CAS Number: 4045-44-7
EINECS: 223-752-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3920 to 1.3940 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR