1-Ethylpiperidine 99%

Supplier: Thermo Scientific Chemicals
Danger

11888-0100 11888-5000
TS11888-0100EA 658 USD
TS11888-0100 TS11888-5000
1-Ethylpiperidine 99%
1-Ethylpiperidine
Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 129…131 °C (760 mmHg)
Melting Pt: –20 °C
Density: 0.824 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00006507
CAS Number: 766-09-6
EINECS: 212-161-6
UN: 2386
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR